1230 Series Stainless Steel Micrometers, L.S. Starrett
Supplier: Safety & Industrial Supplies
681-53197
300045-374EA 463.59 USD
300045-374
1230 Series Stainless Steel Micrometers, L.S. Starrett
Micrometers
The combination ratchet and speeder for uniform pressure and quicker adjustment. Balanced frame and thimble design insure easy handling and better readability.
  • Satin finish stainless steel - no glare - rust and stain resistant
  • Advanced sleeve design with staggered lines and distinct figures for precise and easy readability
  • Convenient decimal equivalents on Inch tools
  • Ring-type knurled lock nut for quick and sure locking
  • Rigid one-piece frame of drop forged steel
  • Carbide measuring faces
Cylindrical anvil tip