Series 531 Precision Vernier Calipers, Mitutoyo
Supplier: Safety & Industrial Supplies
504-531-129
300045-118EA 297.48 USD
300045-118
Series 531 Precision Vernier Calipers, Mitutoyo
Calipers
  • The jaws move only when spring loaded thumb clamp is depressed
  • The finest of vernier calipers for accurate work, with Mitutoyo's exclusive "Quardi" 4-way measuring feature
  • Models available with a fine-adjustment carriage to fine-feed the slider