End Mounted Dial Test Indicators, Brown and Sharpe Precision, ORS Nasco
Supplier: Safety & Industrial Supplies
137-599-7037-3
300044-506EA 886.2 USD
300044-506
End Mounted Dial Test Indicators, Brown and Sharpe Precision, ORS Nasco
Levels
Balanced dial display with 0.0005" graduations.
  • Accurate to ±0.0005"
  • Measurements provided in inches
  • Ball point tip
Delivery information: Indicators with end mounted dials are supplied with contact points, universal dovetail mounting attachment, a contact point wrench, and a certificate of accuracy and traceability in a fitted case.