Weiler® Single Row Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Supplier: Safety & Industrial Supplies
804-13258 804-12306 804-12316 804-12416 804-12746 804-12856 804-12886 804-12356 804-12396 804-12476 804-12816 804-13150 804-13256 804-13281 804-12376P 804-12456 804-13286P 804-12206 804-13286 804-13163 804-13284 804-12276 804-13153 804-13253 804-13025 804-13156 804-12376 804-12846 804-12866 804-12256 804-12326 804-12406 804-12736 804-13160 804-13015 804-13021 804-13257 804-13283 804-13285 804-12316P
300001-006EA 72.33 USD
300001-006 300010-160 300010-161 300010-163 300010-164 300042-560 300042-564 300042-642 300042-646 300042-650 300042-652 300044-120 300044-598 300044-602 300042-644 300042-648 300045-328 300043-344 300001-005 300010-168 300010-172 300042-932 300044-124 300044-596 300001-004 300001-007 300010-162 300042-558 300042-562 300042-930 300043-346 300043-348 300044-118 300044-122 300044-128 300044-572 300044-574 300044-600 300044-604 300044-606 300049-338
Weiler® Single Row Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Brushes
Knot wire cup brush provides heavy-duty, aggressive brushing action, and are best suited for demanding cleaning applications on flat surfaces
These brushes are designed for use with right-angled grinders to remove rust, paint, oxides, and scale.
An ideal tool for removing surface rust and light corrosion from bevel ends prior to welding.