Bleach Sprayers, Chapin™
Supplier: Safety & Industrial Supplies
139-20075
300037-472EA 85.91 USD
300037-472
Bleach Sprayers, Chapin™
Spray Guns
Perfect for remediation and cleaning.
  • Funnel mouth opening for easy and safe filling/cleaning
  • Seals and gaskets designed to tolerate bleach solutions
  • Spray handle with lock-on spraying feature
  • Adjustable poly nozzle
Polyethylene body with plastic handle