Proto® Protoblack™ Pear Head Ratchets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-5449-14BL
300031-236EA 119.87 USD
300031-236
Proto® Protoblack™ Pear Head Ratchets, ORS Nasco
Tool Sets
Unique floating pawl design provides superior strength and long life.
  • Reversing lever designed for quick, one-handed operation
  • Thin head design for access into tight work areas
Certifications: ASME B107.10M.