Torqueplus™ Protoblack™ 12-Point Combination Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
577-1212BASD 577-1256B 577-1216BASD 577-1220BASD 577-1222BASD 577-1226BASD 577-1228BASD 577-1246B 577-1258B 577-1234BASD 577-1240BASD 577-1242B 577-1252B 577-1254B 577-1260B 577-1264B 577-1214BASD 577-1218BASD 577-1224BASD 577-1230BASD 577-1236BASD 577-1244B 577-1248B
300023-614EA 24.02 USD
300023-614 300023-802 300030-776 300030-780 300030-782 300030-786 300030-788 300030-802 300023-806 300030-794 300030-798 300023-788 300023-794 300023-798 300023-810 300023-812 300030-774 300030-778 300030-784 300030-790 300030-796 300030-800 300030-804
Torqueplus™ Protoblack™ 12-Point Combination Wrenches, Proto®, ORS Nasco
Wrenches
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Improves grip on worn, rounded fasteners
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
  • For access to areas where sockets and attachments will not fit or where greater leverage is required