Punch and Die Set, English Measurements, with Plastic Case, ORS Nasco
Supplier: Safety & Industrial Supplies
605-40105
300029-707EA 434.32 USD
300029-707
Punch and Die Set, English Measurements, with Plastic Case, ORS Nasco
Tool Sets
An economical way to make washers, shims, and gaskets.
  • Cut steel, brass, copper, aluminum, rubber, and plastic up to 0.025" thick
  • Cut stainless steel and blue tempered shim up to 0.010" thick
  • Clear plastic top plate is marked with accurate center lines
  • Mark center lines, align with center lines, and hit with hammer