Proto® ProtoGrip™ Hi-Strength Magnetic Impact Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-6908MHF 577-6916HF 577-7217MHF 577-7218HF 577-7416PF 577-7416HF 577-6914HF 577-6910HF 577-6911MPF 577-6914PF 577-7208MHF 577-7211MHF 577-7213MHF 577-7215MHF 577-7414PF
300027-412EA 24.23 USD
300027-412 300027-452 300027-600 300031-354 300031-378 300031-380 300031-446 300031-448 300031-442 300027-876 300027-444 300031-322 300031-328 300031-334 300031-358 300031-366 300031-370 300031-374 300031-444
Proto® ProtoGrip™ Hi-Strength Magnetic Impact Sockets, ORS Nasco
Tool Sets
6-point magnetic sockets use high strength magnets made from a rare earth magnet alloy/nylon blend
  • ProtoGrip™ Magnetic Sockets require no bushings to hold the magnet in place
  • Allows room for more magnet material, helping increase total magnetic strength
  • Designed to easily pick up heavy fasteners and hold them tightly during use
  • Impact sockets with high impact strength are used in automated and hand impact socket application
  • Sockets are available with either inch or metric measurements
Strong magnetic hold helps increase efficiency when installing fasteners in awkward or hard-to-reach locations.