Proto® TorquePlus™ Metric Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-5410M 577-5410MH 577-5411MH 577-5412MH 577-5413M 577-5414M 577-5414MH 577-5415M 577-5418MH 577-5419M 577-5420MH 577-5421MH 577-5422M 577-5422MH 577-5426M 577-5428M 577-5428MH 577-5438M 577-5519M 577-5524M 577-5526M 577-5527M 577-5529M 577-5530M 577-5535M 577-5542M 577-5546M 577-5558M 577-5431M 577-5431MH 577-5425MH 577-5427M 577-5413MH 577-5418M 577-5424M 577-5521M 577-5411M 577-5416MH 577-5426MH 577-5520M 577-5432M 577-5412M 577-5415MH 577-5416M 577-5417M 577-5417MH 577-5419MH 577-5420M 577-5421M 577-5423M 577-5423MH 577-5424MH 577-5425M 577-5427MH 577-5429M 577-5429MH 577-5522M 577-5523M 577-5525M 577-5528M 577-5531M 577-5532M 577-5536M 577-5538M 577-5541M 577-5550M 577-5554M 577-5555M 577-5560M 577-5430M 577-5430MH 577-5432MH 577-5436M
300026-622EA 15.51 USD
300026-622 300026-624 300026-638 300026-652 300026-660 300026-670 300026-672 300026-676 300026-704 300026-708 300026-732 300026-750 300026-758 300026-760 300026-792 300026-808 300026-810 300026-852 300026-962 300026-978 300026-984 300026-986 300026-994 300027-000 300027-014 300027-034 300027-044 300027-068 300031-218 300031-220 300026-784 300026-798 300026-662 300026-702 300026-774 300026-970 300026-636 300026-688 300026-794 300026-968 300031-222 300026-650 300026-678 300026-686 300026-692 300026-694 300026-710 300026-730 300026-748 300026-764 300026-766 300026-776 300026-782 300026-800 300026-814 300026-816 300026-972 300026-974 300026-980 300026-992 300027-002 300027-008 300027-020 300027-024 300027-030 300027-052 300027-060 300027-062 300027-072 300031-214 300031-216 300031-224 300031-228
Proto® TorquePlus™ Metric Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in metric.