Proto® TorquePlus™ Protoblack™ Deep Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-5014B 577-5028B 577-5030B 577-5316B 577-5318B 577-5320B 577-5326B 577-5322B 577-5324B 577-5016B 577-5012B 577-5018B 577-5020B 577-5022B 577-5024B 577-5026B 577-5032B 577-5328B 577-5330B 577-5332B 577-5334B 577-5336B 577-5340B
300025-916EA 15.22 USD
300025-916 300026-000 300026-010 300026-404 300026-414 300026-426 300026-452 300026-436 300026-444 300025-930 300025-902 300025-942 300025-958 300025-970 300025-984 300025-994 300026-016 300026-464 300026-472 300026-482 300026-492 300026-498 300026-516
Proto® TorquePlus™ Protoblack™ Deep Sockets, ORS Nasco
Applies more torque to fastener.
  • Concentric openings
  • Reduces risk of fastener rounding
  • High strength, thin wall design
  • Hand use only
  • Measurments in inches