Proto® TorquePlus™ Deep Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-5330 577-5312 577-5314H 577-5318H 577-5320H 577-5326 577-5328H 577-5332 577-5332H 577-5334 577-5340H 577-5528HL 577-5532HL 577-5540HL 577-5548HL 577-5324H 577-5338 577-5530HL 577-5534HL 577-5324 577-5314 577-5328 577-5342 577-5342H 577-5348H 577-5536HL 577-5330H 577-5336 577-5340 577-5312H 577-5316 577-5316H 577-5318 577-5320 577-5322 577-5322H 577-5326H 577-5334H 577-5336H 577-5338H 577-5344 577-5344H 577-5346 577-5346H 577-5348 577-5542HL 577-5544HL 577-5546HL 577-5552HL
300002-952EA 31.54 USD
300002-952 300026-386 300026-396 300026-416 300026-428 300026-450 300026-466 300026-480 300026-486 300026-490 300026-522 300026-990 300027-006 300027-028 300027-048 300002-950 300026-508 300026-998 300027-012 300002-948 300026-394 300026-462 300026-524 300026-530 300026-554 300027-018 300002-954 300002-956 300002-958 300026-388 300026-402 300026-406 300026-412 300026-422 300026-434 300026-438 300026-454 300026-496 300026-504 300026-514 300026-532 300026-538 300026-540 300026-546 300026-548 300027-032 300027-038 300027-042 300027-056
Proto® TorquePlus™ Deep Sockets, ORS Nasco
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches.