Proto® TorquePlus™ Metric Deep Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-5313M 577-5319M 577-5324M 577-5330M 577-5332M 577-5314M 577-5315M 577-5327M 577-5311M 577-5329M 577-5316M 577-5318M 577-5323M 577-5331M 577-5310M 577-5312M 577-5317M 577-5320M 577-5321M 577-5322M 577-5325M 577-5328M
300026-392EA 26.44 USD
300026-392 300026-420 300026-446 300026-476 300026-488 300026-398 300026-400 300026-460 300026-384 300026-470 300026-408 300026-418 300026-442 300026-478 300026-382 300026-390 300026-410 300026-430 300026-432 300026-440 300026-448 300026-468
Proto® TorquePlus™ Metric Deep Sockets, ORS Nasco
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in metric.