Proto® TorquePlus™ Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-4706 577-4707 577-4708T 577-4709T 577-4710S 577-4710T 577-4712S 577-4714 577-5208H 577-5208S 577-5210 577-5212S 577-5214 577-5214S 577-5216 577-5216S 577-5224 577-5230H 577-5232H 577-25036 577-25038 577-25044 577-25052 577-25054 577-25074 577-25080 577-25084 577-4707T 577-5218 577-5226 577-5232 577-25064 577-25072 577-4712T 577-4708 577-4711 577-4714T 577-4718T 577-5212 577-5218H 577-5222H 577-5228 577-25042 577-25047 577-25058 577-25060 577-25066 577-25070 577-4704 577-4705 577-4706T 577-4708S 577-4709 577-4710 577-4711T 577-4712 577-4716T 577-4718 577-5206H 577-5208 577-5210H 577-5210S 577-5211 577-5212H 577-5214H 577-5216H 577-5220 577-5224H 577-5228H 577-5230 577-25032 577-25034 577-25048 577-25056 577-25062 577-25068 577-25086 577-25088 577-25092 577-25098 577-25104 577-25122 577-5220H 577-5221MH 577-5222 577-5225MH
300025-304EA 9.79 USD
300025-304 300025-318 300025-344 300025-358 300025-388 300025-390 300025-428 300025-440 300026-060 300026-066 300026-074 300026-126 300026-140 300026-154 300026-160 300026-170 300026-238 300026-270 300026-276 300030-844 300030-846 300030-850 300030-856 300030-858 300030-878 300030-880 300030-882 300030-886 300030-892 300030-948 300025-328 300026-176 300026-250 300026-272 300030-868 300030-876 300025-430 300025-332 300025-394 300025-450 300025-470 300026-106 300026-180 300026-232 300026-258 300030-848 300030-852 300030-862 300030-864 300030-870 300030-874 300030-884 300025-280 300025-290 300025-314 300025-342 300025-348 300025-364 300025-412 300025-416 300025-462 300025-464 300026-046 300026-056 300026-084 300026-090 300026-092 300026-120 300026-148 300026-164 300026-190 300026-242 300026-262 300026-264 300030-840 300030-842 300030-854 300030-860 300030-866 300030-872 300030-888 300030-890 300030-894 300030-896 300030-898 300030-950 300030-952 300031-130 300031-138 300031-140 300031-148
Proto® TorquePlus™ Sockets, ORS Nasco
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches (and metric on select models.)
Certifications: ASME B107.5M.