Proto® TorquePlus™ Metric Deep Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
577-4706LM 577-4708LM 577-4710LM 577-4714LM 577-5012MH 577-5013M 577-5014MH 577-5015MH 577-5016MH 577-5017M 577-5018MH 577-5019MH 577-5022M 577-5023M 577-5009MH 577-5010M 577-5020M 577-4704LM 577-47055LM 577-5007M 577-5008MH 577-5011MH 577-5010MH 577-5017MH 577-5020MH 577-5021M 577-4705LM 577-4707LM 577-4709LM 577-4712LM 577-4713LM 577-5006MH 577-5007MH 577-5008M 577-5011M 577-5012M 577-5013MH 577-5014M 577-5015M 577-5016M 577-5018M 577-5019M 577-5021MH 577-5022MH 577-5023MH 577-5009M
300025-308EA 16.71 USD
300025-308 300025-336 300025-382 300025-444 300025-908 300025-910 300025-922 300025-926 300025-936 300025-938 300025-948 300025-952 300025-974 300025-978 300031-088 300091-620 300091-622 300025-286 300025-292 300025-874 300025-884 300025-898 300025-894 300025-940 300025-962 300025-964 300025-298 300025-322 300025-352 300025-422 300025-434 300025-872 300025-876 300025-882 300025-896 300025-906 300025-912 300025-920 300025-924 300025-934 300025-946 300025-950 300025-966 300025-976 300025-980 300031-086
Proto® TorquePlus™ Metric Deep Sockets, ORS Nasco
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in metric.