Torqueplus™ 12-Point Short Combination Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
577-1220TF
300023-714EA 27.38 USD
300023-714 300023-576 300023-732
Torqueplus™ 12-Point Short Combination Wrenches, Proto®, ORS Nasco
Wrenches
Applies more torque to fastener.
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Reduces risk of fastener rounding
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Unique arc design increases surface contact