Torqueplus™ 12-Point Short Box Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
577-1120 577-1122 577-1131
300023-392EA 29.27 USD
300023-392 300023-396 300023-402 300023-412
Torqueplus™ 12-Point Short Box Wrenches, Proto®, ORS Nasco
Wrenches
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Applies more torque to fastener
  • Reduces risk of fastener rounding
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury