Trulock™ Narrow-Face Crimped Wire Wheels, Weiler®
Supplier: Safety & Industrial Supplies
804-01175
300010-123EA 60.42 USD
300010-123
Trulock™ Narrow-Face Crimped Wire Wheels, Weiler®
Abrasives
Narrow-face crimped wire wheels.
  • Flexible brushing action
  • Can be mounted in multiples
  • Steel wires
Use these wire wheels for cleaning rust, scale and dirt, light de-burring, edge blending, roughening for adhesion, and finish preparation prior to plating or painting.