Lava® Hand Cleaners, WD-40
Supplier: Safety & Industrial Supplies
780-10185
300010-106CS 57.99 USD
300010-106
Lava® Hand Cleaners, WD-40
Skincare Soaps and Skin Cleansers
Hand cleanser removes grease, adhesive, caulk, dust, oil, paint, tar.
  • Moisturizes
  • Pumice is used in combination with other cleaning agents
  • 10.30 pH
Weight: 0.46 lbs.