Powerlock® Tape Rules, ³/₄" Wide Blade, Stanley® Products
Supplier: Safety & Industrial Supplies
680-33-116
300009-898EA 78.37 USD
300009-898
Powerlock® Tape Rules, ³/₄" Wide Blade, Stanley® Products
Measuring Tapes
When taking measurements, the secure blade lock will not creep.
  • 7' blade standout
  • Mylar® polyester film extends life of entire blade
  • Chrome high-impact ABS case is job site tough
  • Corrosion-resistant Tru-zero end hook for accurate measurements
Single scale, with measurements in inches.