Torpedo Levels, Ridgid®
Supplier: Safety & Industrial Supplies
632-20233
300009-754EA 55.39 USD
300009-754
Torpedo Levels, Ridgid®
Levels
Glow surround view for easier reading of vials in dark or shadowed areas.
Rare-earth magnets five times stronger than standard strip magnets.