Temflex™ Vinyl Electrical Tape 1700, ORS Nasco, INC.
Supplier: Safety & Industrial Supplies
405-076308-92568
300009-347CS 272.98 USD
300009-347
Temflex™ Vinyl Electrical Tape 1700, ORS Nasco, INC.
Tapes
Primary electrical insulation for all wire and cable splices rated up to 600 volts.
  • Polyvinyl chloride (PVC) tape is flame-retardent and conformable
  • Provides excellent mechanical protection with minimum bulk