No. 410 & 411 Lightweight Xenoy Safety Lockout Padlocks, Master Lock
Supplier: Safety & Industrial Supplies
470-410RED 470-411RED
300009-177PK 108.9 USD
300009-177 300009-179
No. 410 & 411 Lightweight Xenoy Safety Lockout Padlocks, Master Lock
Designed by safety directors specifically for OSHA lockout compliance - 1 3/4" tall.
  • Durable, lightweight, non-conductive xenoy lock body
  • Customize on-site with permanent, write-on labels
  • High security, reserved-for-safety cylinder - Up to 100,000 key changes
  • Key retaining - Ensures that padlocks are not left unlocked
  • Eight Colors Available - Add Desired Suffix - RED, YLW, BLU, GRN, BLK, ORJ, PRP, TEAL
Certifications: Compliance with OSHA " One Employee, One Lock, One Key" directive