ProLine™ Carbon Dioxide Fire Extinguishers - BC Type, Kidde
Supplier: Safety & Industrial Supplies
408-466180
300008-559EA 318.37 USD
300008-559
ProLine™ Carbon Dioxide Fire Extinguishers - BC Type, Kidde
First Aid Supplies Fire Extinguishers
The agent of this carbon dioxide fire extinguisher BC will not contaminate food, valuable materials or electronics. It extinguishes electrical equipment, flammable liquids, gases and solvents.
  • Seamless cylinder is strong and lightweight
  • Clean agent
  • CO₂ gas dissipates quickly without leaving residue
HxØ: 17x5.25"
Charge weight: 5 lbs.
Unit weight: 15 lbs.