SR 450 Series Single Stage Heavy Duty Regulators, Victor
Supplier: Safety & Industrial Supplies
341-0781-0527
300008-239EA 642.25 USD
300008-239
SR 450 Series Single Stage Heavy Duty Regulators, Victor
Welding Supplies
  • Medium to high gas service capacity
  • Single stage design
  • Stem type seat mechanism
  • Delrin® cap bushing for smooth adjustments
  • External self reseating relief valve not designed to protect downstream apparatus