Economy Balloon Inflators with Gauge, Western Enterprises
Supplier: Safety & Industrial Supplies
312-R125G
300008-007EA 87.14 USD
300008-007
Economy Balloon Inflators with Gauge, Western Enterprises
Welding Supplies
  • Horizontal one-piece design with all brass piston construction
  • Replaceable Viton® O-ring
  • Hand tight design provides leak-free connection
  • Replaceable rubber tilt valve
  • Easy-to-read contents gauge with rubber protective cover