The Shield* Safety Goggles, Jackson, Kimberly-Clark
Supplier: Safety & Industrial Supplies
138-3010343
300007-063EA 49.74 USD
300007-063
The Shield* Safety Goggles, Jackson, Kimberly-Clark
Goggles
Cylindrical shaped anti-fog lens made of polycarbonate offers greater range of visibility.
  • Provides extreme visual clarity in all conditions
  • Superior impact and chemical splash protection
  • Superior eye protection from 99.9% of UV radiation
  • Full face protection with detachable faceshield
  • Indirect ventilation
  • Polycarbonate curved shield conforms to face, offering increased facial protection
Weight: 0.46 lbs.