Blade Disposal Containers, Stanley®
Supplier: Safety & Industrial Supplies
680-11-080
300003-892EA 49.76 USD
300003-892
Blade Disposal Containers, Stanley®
Bins Sharps Containers
For utility-knife blades and single-edged razor blades only.
  • High impact, puncture-resistant container
  • Snap-on lid locks tight for safety and is tethered to the container