Proto® Drift Punch Set, ORS Nasco
Supplier: Safety & Industrial Supplies
577-96A
300003-226PK 193.98 USD
300003-226
Proto® Drift Punch Set, ORS Nasco
Tool Sets
Made from S2 steel and specially heat-treated for greater strength and durability, with a modified parabolic striking end to help reduce mushrooming and chipping.
  • Reverse taper to help prevent binding
  • Black oxide finish to help prevent rust corrosion; extending tool life and performance
  • English measurements
  • Packaged 7 per kit pouch
A polished identification panel on one of the flats is marked with the tool number and size in a black inlay. Set offers a wide range of sizes to handle a variety of jobs. Includes a pouch for convenient storage.