Proto® Power Track ll™ Ergonomics™ Tongue and Groove Pliers, ORS Nasco
Supplier: Safety & Industrial Supplies
577-260SG 577-264SG 577-265SG 577-261SG
300024-228EA 82.64 USD
300024-228 300024-276 300024-328 300002-838 300024-246
Proto® Power Track ll™ Ergonomics™ Tongue and Groove Pliers, ORS Nasco
Pliers
Pliers features precision milled undercut groove joint.
  • Plastisol grips for added comfort
  • Easily adjusts
  • Straight, serrated jaws
  • Forged undercut
Certifications: Federal Spec. GGG-P-471e.