Proto® Long Needle Nose Plier, ORS Nasco
Supplier: Safety & Industrial Supplies
577-228G
300002-820EA 50.15 USD
300002-820
Proto® Long Needle Nose Plier, ORS Nasco
Pliers
Diamond serrated jaw.
  • Plastisol grips for added comfort
  • Long handle provides added leverage
Certifications: Federal Spec. GGG-P-471e; ANSI Spec. B107.13M.