660™ Quick Metal® Retaining Compound, Loctite, Henkel
Supplier: Safety & Industrial Supplies
442-66040
300001-034EA 67.48 USD
300001-034
660™ Quick Metal® Retaining Compound, Loctite, Henkel
Adhesives
A creamy, non-running adhesive/sealant with mild odor.
  • Repairs worn machine parts
  • Restores correct fit to mating assemblies
  • Acrylic urethane as chemical compound
It is resistant to ethanol, motor oil and unleaded petrol and applicable for most metals and joint.