Dykem™ Brite-Mark™ Medium Markers, ORS Nasco
Supplier: Safety & Industrial Supplies
253-84002
300000-769PK 80.01 USD
300000-769
Dykem™ Brite-Mark™ Medium Markers, ORS Nasco
Pens Permanent Markers
Bullet tips are replaceable to assure complete use of paint in each marker
  • Drying time is 25 – 35 seconds
  • Heavy-duty barrel retains its shape
  • Ideal for color coding
Makes permanent bright marks on metal, rubber, plastic, glass, concrete, leather, stone, wood, vinyl, and more.
Valve action mechanism prevents paint in barrel from drying when cap is left off.