Mr. Clean® Finished Floor Cleaners, Procter & Gamble
Supplier: Safety & Industrial Supplies
608-02621
300000-754PK 235.24 USD
300000-754
Mr. Clean® Finished Floor Cleaners, Procter & Gamble
Cleaning Agents Detergents and Surface Cleansers
Low-sudsing formula is suitable for automatic floor scrubbing machines.
  • Patented formula removes dirt while protecting the finish
  • No-rinse cleaner leaves no dulling residue and minimizes yellowing
  • Recommended dilution: manual (1:128), automatic machines (1:512)
  • Lemon scent