Economy Balloon Inflators, Western Enterprises
Supplier: Safety & Industrial Supplies
312-R125
300000-072EA 63.77 USD
300000-072
Economy Balloon Inflators, Western Enterprises
Welding Supplies
  • Horizontal one-piece design with all brass piston construction
  • Replaceable Viton® O-ring
  • Hand tight design provides leak-free connection
  • Replaceable rubber tilt valve