Nucleosil® HPLC Columns, C18 HD, Macherey Nagel
Supplier: Macherey-Nagel
720193.46
29444-592EA 2813.54 USD
29444-592
Nucleosil® HPLC Columns, C18 HD, Macherey Nagel
Chromatography Columns
These octadecyl modified high density silica phase for HPLC have a monomeric modification and are encapped.
  • Hydrophobic, nonpolar phase for RP chromatography
  • Octadecyl modified, high density silica phase
These octadecyl modified, high density silica phase for HPLC have a monomeric modification and are encapped. pH stability 2 to 9, carbon content 20%. Suited for separation of basic compounds such as pharmaceuticals.