CHROMABOND® XTR, SPE Columns, Macherey-Nagel
Supplier: Macherey-Nagel
730507
29444-586PK 392.16 USD
29444-586
CHROMABOND® XTR, SPE Columns, Macherey-Nagel
Chromatography Columns
SPE columns, CHROMABOND® XTR, coarse-grained kieselguhr base material with large pore size, high pore volume and constantly high batch-to-batch quality.
  • Fast, reproducible and economical
  • Simultaneous preparation of several samples
  • No problems with phase separation and formation of emulsions
  • High recovery rates, saving of time and solvents
  • Organic solutions need not to be dried after separation
Suitable for liquid-liquid extraction of highly viscous aqueous solutions such as physiological fluids (blood, plasma, and serum) in clinical chemistry, dyes in textiles, environmental and food analysis without use of a separation funnel. High water loadability without breakthrough of water during elution with organic solvents. Also suited for removing small amounts of water from solvents which are not miscible with water.