TLC Plates, Silica Gel 60, Unmodified Layers, SIL G, Macherey-Nagel
Supplier: Macherey-Nagel
809013
29444-516PK 612.35 USD
29444-516
TLC Plates, Silica Gel 60, Unmodified Layers, SIL G, Macherey-Nagel
TLC Plates
Unmodified standard silica layers for TLC.
  • Layer thickness for analytical plates 0.25 mm and preparative plates 0.5 and 1 mm
  • High quality silica gel
  • Specific surface (BET) ~500 m2/g
  • Particle size 5 to 17 μm
  • Binders are stable in almost all organic solvents
Indicators like manganese helps in activating zinc silicate with green fluorescence in short-wave UV light (254 nm) and special inorganic luminescent pigment with blue fluorescence in long-wave UV light (366 nm).
SIL G glass plates with a layer thickness from 500 to 2000 µm are used for preparative TLC. SIL G-25 HR plates are coated with high purity silica 60, bonded with gypsum and a very small quantity of a polymeric organic binder. They are recommended for aflatoxin separations. SIL G-25 Tenside plates are impregnated with ammonium sulphate and are applied for separations of surfactants.
These highly robust plates and flexible sheets with gypsum and polymeric binder systems are well-suitable for every standard application.