Corning® Pipette Tips, Microvolume
Supplier: Corning
4826 4830 4834 4840 4894 4901
29443-298CS 91.6 USD
29443-298 29443-300 29443-302 29443-304 29443-320 29443-322
Corning® Pipette Tips, Microvolume
Pipette Tips
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
  • All racked tips are certified RNase-/DNase-free and non pyrogenic
  • Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles