Costar® 60 mm Center Well Culture Dish, Corning
Supplier: Corning
3260
29442-026CS 2681.93 USD
29442-026
Costar® 60 mm Center Well Culture Dish, Corning
Culture Dishes Cell Culture Dishes
Optically clear PS IVF culture dish with 20 mm centre well. Inner well holds 3 ml of medium while the outer well holds 10 ml.
  • 45° slope funnels oocyte/embryo into centre of well
  • Stand-off rim on base protects optical surface from scratching
  • Treated for optimal cell attachment
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
Product is strictly not for human use. The center well dish is a Class I medical device and should never be used in applications which require the product to be certified as a Class II or Class III medical device. This product is not CE marked.
Center well beveled 'lead in' makes it easier to manipulate a pipette. Fill center well to the bottom of bevel for 1 ml volume. Dish cover has edge beads permit uniform gas flow. Outer moat provides both humidification and washing.
For research use only.
Ordering information: Accessories not included.