Minimate™ Tangential Flow Filtration Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
OA001C12 OA003C12 OA005C12 OA010C12 OA050C12 OA070C12 OA500C12 OA990C12 OA030C12 OA100C12 OA300C12
29301-900PK 304 USD
29301-900 29301-902 29301-904 29301-908 29301-912 29301-914 29301-922 29301-924 29301-910 29301-916 29301-920
Minimate™ Tangential Flow Filtration Capsules, Cytiva (Formerly Pall Lab)
Filters Tangential Flow Cassettes
The disposable Minimate™ TFF capsules, with Omega™ membrane, can perform concentration processes desalting or buffer exchange on the same system with minimal user intervention, and achieves high concentration factors in a single step due to the low hold-up volume of the devices.
  • Scaleable
  • Works with a variety of positive pressure laboratory pumps
  • High product recovery
  • 100% integrity tested
The capsules will process samples of up to 1 litre, or efficiently concentrate samples as little as 5 ml.
Delivery information: Supplied with all fittings andMore Product Information