Nalgene® Fernbach Culture Flask, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
4105-2800
29171-854CS 493.39 USD
29171-854
Nalgene® Fernbach Culture Flask, Polycarbonate, Thermo Scientific
Flasks Cell Culture Flasks
For growing large-scale cultures
Window-clear polycarbonate construction. Break-resistant. Autoclavable, but repeated autoclaving may reduce mechanical strength.