KIMAX® Pickel Extraction Flask, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
26850-99
29140-607CS 340.19 USD
29140-607
KIMAX® Pickel Extraction Flask, Kimble Chase
Flasks Flat Bottom Flasks
KIMAX® flask with three indentations spaced 120 degrees apart
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The diameter at top is closely controlled to insure a fit with metal condensers.