PYREX® Flasks, Flat Bottom, Glass, Low Actinic, with Standard Tapered Joints, Corning
Supplier: Corning
54100-250
29113-148CS 154.55 USD
29113-148
PYREX® Flasks, Flat Bottom, Glass, Low Actinic, with Standard Tapered Joints, Corning
Flasks Flat Bottom Flasks
PYREX® low actinic glass, short neck.
  • High mechanical strength
  • Resistant to chemicals
  • Maximum working temperature: 230 °C (446 °F)
These flasks have full length outer standarrd tapered 24/40 joint. The normal working temperature is 230 °C. Low actinic stained glass provides protection for materials sensitive to light.