Aspirator Filter Pumps, Humboldt Manufacturing
Supplier: Humboldt Manufacturing
H-12100 H-12020
28651-005EA 313.38 USD
28651-005 89370-258 89370-278
Aspirator Filter Pumps, Humboldt Manufacturing
Pumps Vacuum Aspiration Systems
Humboldt Aspirator Filter Pumps offer excellent performance for vacuum filtrations under both high and low water pressure conditions
The pumps feature a ball-type, positive-action valve in the suction tube which prevents water backflow if the pressure is reduced.