Nalgene® Polypropylene Vacuum Pump Aspirator, Thermo Scientific
Supplier: Thermo Fisher Scientific
6140-0010
28610-008CS 674.46 USD
28610-008
Nalgene® Polypropylene Vacuum Pump Aspirator, Thermo Scientific
Pumps Vacuum Aspiration Systems
Maximum free air pumping capacity of 11.5L per minute at a water flow rate of 6.5L per minute
Provides vacuum to 723.9mm Hg (28.5in. Hg). Operates efficiently at water pressures as low as 7.5psig. Safe with corrosive materials. (NNI 6140-0010)
Ordering information: With integral check valve, male 0.95cm (3/8") NPT connecting thread, and 12.1cm (43/4") length of polyethylene tubing for discharge end.