Whatman™ Grade GF/C Glass Microfiber Filters, Binder Free
Supplier: Cytiva
1822-9916 1822-021 1822-024 1822-025 1822-042 1822-047 1822-055 1822-070 1822-090 1822-110 1822-125 1822-150 1822-100 1822-037 1822-050 1822-185 1822-6580 1822-849 1822-866
14005-870EA 484.52 USD
14005-870 28497-620 28497-641 28497-652 28497-685 28497-696 28497-700 28497-721 28497-743 28497-765 28497-776 28497-787 28497-619 28497-754 28497-663 10035-304 10035-306 10035-308 10035-310 10035-312 10035-314
Whatman™ Grade GF/C Glass Microfiber Filters, Binder Free
Filters Glass Fiber Filters
These borosilicate glass filters are widely used as total suspended solids filters for applications testing both potable water and waste water. GF/C can also function as a filter for liquid scintillation counting, cell harvesting, industrial waste analysis and binding assays that require high loading capacity.
  • Fine particle retention
  • Good flow rate and loading capacity
  • Binder free