Whatman™ Grade 802 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
5802-150 5802-240 5802-320 5802-125 5802-185 5802-385 5802-6698
28467-606PK 61.67 USD
28467-606 28467-607 28467-611 70240-110 70240-112 70240-114 10035-482
Whatman™ Grade 802 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A prepleated filter for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
  • Highest retention among standard filters
  • Slow flow rate
  • Fine-particle retention
  • For qualitative analysis
The filter is wet-strengthened and for normal qualitative application it will not introduce any significant impurities into the filtrate. However, it is not recommended for Kjeldahl nitrogen analysis.