Whatman™ Grade 2V Qualitative Filter Papers, Fluted, Whatman products (Cytiva)
Supplier: Cytiva
1202-125 1202-150 1202-185 1202-240 1202-320 1202-385 1202-400 1202-270 1202-500
28465-022PK 120.22 USD
28465-022 28465-044 28465-066 28465-088 28465-102 28465-124 70240-038 10035-256 10147-912
Whatman™ Grade 2V Qualitative Filter Papers, Fluted, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Compared with quadrant-folded filters, Cytiva's Whatman Grade 2V pre-folded filter paper exposes extra surface area, which enables a higher loading capacity and shorter filtration time.
  • Medium flow and nominal particle retention rating of 8 µm
  • Herzberg filtration speed of 240s
  • Fluted (pre-pleated) for easy use with conical filter funnels
  • Can be used for soil analysis or general filtration
Grade 2V pre-folded filter sheets are suitable for extracting samples from soil to support contaminant testing in the presence of weakly acidic solutions. Pre-folded filters for seed and soil testing.