CultureGard® HF Perfusion Filter, Spectrum® Laboratories
Supplier: Spectrum Laboratories
CU2M-205-12N
28196-400PK 1827.53 USD
28196-400
CultureGard® HF Perfusion Filter, Spectrum® Laboratories
Filters In-Line Filters
The specialized, autoclavable filter utilizes a 0.2 µm DynaFibre® membrane that permits the flow of culture medium while preventing the introduction of foreign microbials
By using two CultureGard® filters in series, users can virtually eliminate the risk of culture contamination during media feeding and culture perfusion. Should the first filter become plugged, simply remove it and reattach the media supply directly to the second filter already in place, maintaining the sterile barrier to the culture. Packaged nonsterile in individual protective pouches.
Potting material: Polyurethane
Suggested volume: 2 to 50 L